Learn More
Aviva Systems Biology Corp Dcaf7 Antibody - N-terminal region (ARP52110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dcaf7. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1700012F10Rik, 2610037L01Rik, C86529, HAN11, Wdr68ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-638