Learn More
Eurofins Discoverx DCAMKL2, active, 10 µg. (14-716)
Supplier:  Eurofins Discoverx 14716
Encompass
N-terminal 6xHis-tagged recombinant, human DCAMKL2 amino acids 376-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9227