Learn More
Medchemexpress LLC HY-134582 5mg , dCBP-1 CAS:2484739-25-3 Purity:>98%
SDP
Catalog No. 502259010
Encompass_Preferred
HY-134582 5mg dCBP-1 CAS: 2484739-25-3 dCBP-1 is a potent and selective heterobifunctional degrader of p300/CBP based on Cereblon ligand. dCBP-1 is exceptionally potent at killing multiple myeloma cells and ablates oncogenic enhancer activity driving MYC expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9010 Supplier Medchemexpress LLC Supplier No. HY1345825MG
HY-134582 5mg dCBP-1 CAS: 2484739-25-3 dCBP-1 is a potent and selective heterobifunctional degrader of p300/CBP based on Cereblon ligand. dCBP-1 is exceptionally potent at killing multiple myeloma cells and ablates oncogenic enhancer activity driving MYC expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.