Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dcc

Catalog No. D800025KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D800025KG 5 kg
D80002100G 100 g
D800021KG 1 kg
D8000225G 25 g
D80002500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. D800025KG Supplier Sigma Aldrich Supplier No. D800025KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dcc

Specifications

CAS 538-75-0
Melting Point 34°C to 35°C
Boiling Point 122°C to 124°C (6 mmHg, lit.)
Molecular Formula C13H22N2
Linear Formula C6H11N=C=NC6H11
Quantity 5 kg
MDL Number MFCD00011659
Synonym N,N'-Dicyclohexylcarbodiimide
Molecular Weight (g/mol) 206.33 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.