Learn More
Medchemexpress LLC HY-136256 5mg Medchemexpress, DCC-3014 CAS:1628606-05-2 Purity:>98%
SDP
Catalog No. 502081429
Encompass_Preferred
Medchemexpress, HY-136256 5mg DCC-3014 CAS:1628606-05-2 DCC-3014 is a c-FMS (CSF-IR) and c-Kit dual inhibitor extracted from patent WO2014145025A2, Compound Example 10, has IC50s of <0.01 μM and 0.1-1 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1429 Supplier Medchemexpress LLC Supplier No. HY1362565MG
Medchemexpress, HY-136256 5mg DCC-3014 CAS:1628606-05-2 DCC-3014 is a c-FMS (CSF-IR) and c-Kit dual inhibitor extracted from patent WO2014145025A2, Compound Example 10, has IC50s of <0.01 μM and 0.1-1 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.