Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dcc-3116 | 2543673-19-2 | 99.7% | 535.60 | 250 MG
SDP

Catalog No. 5000394811
Encompass_Preferred

DCC-3116 is an orally active ULK1/2 inhibitor. It inhibits autophagy in lung cancer cells by targeting KRASG12C signaling, thereby reducing the proliferation of lung cancer cells and exhibiting anti-cancer effects.

  • Orally active ULK1/2 inhibitor
  • Inhibits autophagy in lung cancer cells
  • Targets KRASG12C signaling to reduce cell proliferation
  • Demonstrates anti-cancer effects
  • High purity of 99.7%
  • Consistent with chemical structure by ¹H NMR and MS
  • Stable for 3 years at -20°C in pure form

Catalog No. 50-003-94811 Supplier Medchemexpress LLC Supplier No. 250MGHY160699
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DCC-3116 is an orally active ULK1/2 inhibitor. It inhibits autophagy in lung cancer cells by targeting KRASG12C signaling, thereby reducing the proliferation of lung cancer cells and exhibiting anti-cancer effects.

  • Orally active ULK1/2 inhibitor
  • Inhibits autophagy in lung cancer cells
  • Targets KRASG12C signaling to reduce cell proliferation
  • Demonstrates anti-cancer effects
  • High purity of 99.7%
  • Consistent with chemical structure by ¹H NMR and MS
  • Stable for 3 years at -20°C in pure form

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.