Learn More
Medchemexpress LLC DCC-3116 | 2543673-19-2 | 99.7% | 535.60 | 50 MG
SDP
Catalog No. 5000394420
Encompass_Preferred
DCC-3116 is an orally active ULK1/2 inhibitor. It inhibits autophagy in lung cancer cells by targeting KRASG12C signaling, which in turn inhibits the proliferation of lung cancer cells and exhibits anti-cancer effects.
  • Orally active ULK1/2 inhibitor.
  • Inhibits autophagy in lung cancer cells by inhibiting KRASG12C signaling.
  • Inhibits proliferation of lung cancer cells.
  • Exerts anti-cancer effects.
  • DCC-3116 (100 nM, 72 h) alone inhibits the proliferation of NCI-H2122 and Calu-1 cells, decreases the expression of pS318-ATG13, and shows synergistic effects when combined with Sotorasib.
  • In mouse models of NSCLC lung cancer, DCC-3116 (3 or 30 mg/kg/day for 56 days, p.o. and p.i.) alone or in combination with Sotorasib can inhibit tumor growth and improve survival rate.
Catalog No. 50-003-94420 Supplier Medchemexpress LLC Supplier No. HY16069950MG
Add to Cart
Add to Cart
DCC-3116 is an orally active ULK1/2 inhibitor. It inhibits autophagy in lung cancer cells by targeting KRASG12C signaling, which in turn inhibits the proliferation of lung cancer cells and exhibits anti-cancer effects.
  • Orally active ULK1/2 inhibitor.
  • Inhibits autophagy in lung cancer cells by inhibiting KRASG12C signaling.
  • Inhibits proliferation of lung cancer cells.
  • Exerts anti-cancer effects.
  • DCC-3116 (100 nM, 72 h) alone inhibits the proliferation of NCI-H2122 and Calu-1 cells, decreases the expression of pS318-ATG13, and shows synergistic effects when combined with Sotorasib.
  • In mouse models of NSCLC lung cancer, DCC-3116 (3 or 30 mg/kg/day for 56 days, p.o. and p.i.) alone or in combination with Sotorasib can inhibit tumor growth and improve survival rate.