Learn More
Aviva Systems Biology Corp DCC Antibody - middle region (ARP46583_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46583_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCC. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human DCC. Alternate names: CRC18, CRCR1, IGDCC1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-266