Learn More
AdipoGen DCCH
SDP
Supplier:  AdipoGen CDXD0035M050
Encompass_Preferred
Chemical. CAS 100343-98-4. Formula C14H17N3O3. MW 275.3. Synthetic. Derivatizing agent for carboxylic acid detection. Has also been used for fluorescence detection of monoterpene alcohols.
Catalog No. 50-256-1297