Learn More
Aviva Systems Biology Corp DCDC2 Antibody - middle region (ARP56893_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56893_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCDC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DCDC2. Alternate names: DCDC2A, RU2, RU2SApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-231