Learn More
Aviva Systems Biology Corp Dcdc2a Antibody - N-terminal region (ARP56892_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dcdc2a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Dcdc2, MGC54817, RU2, AW492955ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-230