Learn More
AdipoGen DCFDA, 2044-85-1, MFCD00037501, 1 g
SDP
Catalog No. 501147000
Encompass_Preferred
MF: C24H14Cl2O7, MW: 485.27, ≥95%, Appearance: White to off-white powder. A cell-permeable fluorogenic dye useful for the detection of reactive oxygen species (ROS) and nitric oxide (NO) and for the determination of the degree of overall oxidative stress. The fluorescence of DCFDA indicates oxidation by hydrogen peroxide, peroxynitrite or hydroxyl radical.
Catalog No. 50-114-7000 Supplier AdipoGen Supplier No. CDXD0063G001
MF: C24H14Cl2O7, MW: 485.27, ≥95%, Appearance: White to off-white powder. A cell-permeable fluorogenic dye useful for the detection of reactive oxygen species (ROS) and nitric oxide (NO) and for the determination of the degree of overall oxidative stress. The fluorescence of DCFDA indicates oxidation by hydrogen peroxide, peroxynitrite or hydroxyl radical.