Learn More
Aat Bioquest DCFH-DA [2',7'-Dichlorodihydrofluorescein diacetate] *CAS 4091-99-0*
SDP
Supplier:  Aat Bioquest 15204
Encompass_Preferred
DCFH-DA, 2',7'-Dichlorodihydrofluorescein diacetate (also called 2',7'-dichlorofluorescin diacetate), is hydrolyzed by cellular esterases to 2',7'-dichlorodihydrofluorescein (also called 2',7'-dichlorofluorescin) and is then oxidized to 2',7'-dichlorofluorescein primarily by H2O2.
Catalog No. 50-195-3551