Learn More
Medchemexpress LLC Dcg-iv | 147782-19-2 | MFCD00673968 | >98.0% | 203.15 g/mol | C7H9NO6 | 1 MG
SDP
Catalog No. 5000211507
Encompass_Preferred
DCG-IV is a research chemical used to selectively activate group II metabotropic glutamate receptors (mGlu2 and mGlu3) in neuroscience studies. It is reported to have anticonvulsive and neuroprotective effects and is supplied as a solid for experimental use.
  • Potent agonist of group II mGlu receptors with low-micromolar to sub-micromolar activity.
  • Distinct pharmacological profile useful for dissecting mGlu2/mGlu3 signaling.
  • Reported anticonvulsive and neuroprotective effects in preclinical studies.
  • Supplied as a solid, facilitating storage and formulation for assays.
  • Well characterized chemically (CAS 147782-19-2; formula C7H9NO6; MW 203.15 g/mol).
Catalog No. 50-002-11507 Supplier Medchemexpress LLC Supplier No. HY1013351MG
DCG-IV is a research chemical used to selectively activate group II metabotropic glutamate receptors (mGlu2 and mGlu3) in neuroscience studies. It is reported to have anticonvulsive and neuroprotective effects and is supplied as a solid for experimental use.
  • Potent agonist of group II mGlu receptors with low-micromolar to sub-micromolar activity.
  • Distinct pharmacological profile useful for dissecting mGlu2/mGlu3 signaling.
  • Reported anticonvulsive and neuroprotective effects in preclinical studies.
  • Supplied as a solid, facilitating storage and formulation for assays.
  • Well characterized chemically (CAS 147782-19-2; formula C7H9NO6; MW 203.15 g/mol).