Learn More
Cayman Chemical DchIroInosItol 1234 5 6 10mg
SDP
Catalog No. 502894242
Encompass_Preferred
A stereoisomer of IP6; identified in soil samples from Madagascar, New Zealand, and the Falkland Islands and in freshwater sediment from Danish lakes
Catalog No. 50-289-4242 Supplier Cayman Chemical Supplier No. 900234110 MG
A stereoisomer of IP6; identified in soil samples from Madagascar, New Zealand, and the Falkland Islands and in freshwater sediment from Danish lakes