Learn More
Sigma Aldrich Fine Chemicals Biosciences 4,5-Dicyanoimidazole | ≥98% | Mol. Wt. 96.08 | CAS 6627-59-4 MDL MFCD00133341 | 4 x 2.5L
Catalog No. NC2611335
Encompass
4,5-Dicyanoimidazole (DCI) | Purity: ≥98% | Mol Wt: 96.08 | CAS No: 6627-59-4 | MDL No: MFCD00133341 | 2.5 LDCI is an activator solution used during oligonucleotide synthesis to catalyze the coupling reaction between phosphoramidites and the growing oligonucleotide chain. This product contains 0.5 M DCI dissolved in acetonitrile. It is packaged in 4 x 2.5 L containers.
Catalog No. NC2611335 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L0321004X2.5L
4,5-Dicyanoimidazole (DCI) | Purity: ≥98% | Mol Wt: 96.08 | CAS No: 6627-59-4 | MDL No: MFCD00133341 | 2.5 LDCI is an activator solution used during oligonucleotide synthesis to catalyze the coupling reaction between phosphoramidites and the growing oligonucleotide chain. This product contains 0.5 M DCI dissolved in acetonitrile. It is packaged in 4 x 2.5 L containers.