Learn More
Aviva Systems Biology Corp DCK Antibody - middle region (ARP51775_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51775_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DCK. Alternate names: MGC117410, MGC138632ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60613