Learn More
Selleck Chemical LLC DCLK1-IN-1-5mg
SDP
Supplier:  Selleck Chemical LLC S89765MG
Encompass_Preferred
DCLK1-IN-1 is a selective, in vivo-compatible chemical probe of the doublecortin like kinase 1 (DCLK1) domain that binds to DCLK1 and DCLK2 with IC50 of 9.5 nM and 31 nM, respectively. DCLK1-IN-1 inhibits DCLK1 and DCLK2 kinases with IC50 of 57.2 nM and 103 nM in kinase assay, respectively.
Catalog No. 50-238-9802