Learn More
Aviva Systems Biology Corp DCLRE1C Antibody - C-terminal region (ARP51165_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51165_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCLRE1C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DCLRE1C. Alternate names: A-SCID, DCLREC1C, FLJ11360, FLJ36438, RS-SCID, SCIDA, SNM1C, hSNM1CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60508