Learn More
Aviva Systems Biology Corp DCN Antibody - N-terminal region (ARP48199_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48199_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DCN. Alternate names: CSCD, DSPG2, PG40, PGII, PGS2, SLRR1BApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-59752