Learn More
Medchemexpress LLC HY-108599 1mg Medchemexpress, DCP-LA CAS:28399-31-7 Purity:>98%
SDP
Catalog No. 501968591
Encompass_Preferred
Medchemexpress, HY-108599 1mg DCP-LA CAS:28399-31-7 DCP-LA (FR236924), a linoleic acid derivative, selectively and directly activates PKCε. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8591 Supplier Medchemexpress LLC Supplier No. HY1085991MG
Medchemexpress, HY-108599 1mg DCP-LA CAS:28399-31-7 DCP-LA (FR236924), a linoleic acid derivative, selectively and directly activates PKCε. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.