Learn More
Medchemexpress LLC DCP-LA | 28399-31-7 | 98.0% | 308.50 | 25 MG
SDP
Catalog No. 5000347463
Encompass_Preferred
DCP-LA is a linoleic acid derivative that selectively and directly activates PKCε. It also activates Ca(2+)/calmodulin-dependent protein kinase II (CaMKII) and inhibits protein phosphatase-1 (PP-1) to stimulate AMPA receptor exocytosis. Additionally, DCP-LA inhibits the activation of caspase-3/-9 and helps protect neurons from oxidative stress-induced apoptosis.
  • Selectively and directly activates PKCε
  • Activates Ca(2+)/calmodulin-dependent protein kinase II (CaMKII)
  • Inhibits protein phosphatase-1 (PP-1)
  • Stimulates AMPA receptor exocytosis
  • Inhibits activation of caspase-3/-9
  • Helps protect neurons from oxidative stress-induced apoptosis
  • Facilitates hippocampal synaptic transmission
  • Exhibits protective effect against ischemic brain damage
Catalog No. 50-003-47463 Supplier Medchemexpress LLC Supplier No. 25MGHY108599
DCP-LA is a linoleic acid derivative that selectively and directly activates PKCε. It also activates Ca(2+)/calmodulin-dependent protein kinase II (CaMKII) and inhibits protein phosphatase-1 (PP-1) to stimulate AMPA receptor exocytosis. Additionally, DCP-LA inhibits the activation of caspase-3/-9 and helps protect neurons from oxidative stress-induced apoptosis.
  • Selectively and directly activates PKCε
  • Activates Ca(2+)/calmodulin-dependent protein kinase II (CaMKII)
  • Inhibits protein phosphatase-1 (PP-1)
  • Stimulates AMPA receptor exocytosis
  • Inhibits activation of caspase-3/-9
  • Helps protect neurons from oxidative stress-induced apoptosis
  • Facilitates hippocampal synaptic transmission
  • Exhibits protective effect against ischemic brain damage