Learn More
Aviva Systems Biology Corp DCP1A Antibody - N-terminal region (ARP34582_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34582_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DCP1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DCP1A. Alternate names: SMIF, SMAD4IP1, HSA275986, Nbla00360ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55461