Learn More
Aviva Systems Biology Corp DCP1B Antibody - N-terminal region (ARP50809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCP1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DCP1B. Alternate names: DCP1, hDcp1bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53640