Learn More
Aviva Systems Biology Corp DCP2 Antibody - C-terminal region (ARP41150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DCP2. Alternate names: FLJ33245, NUDT20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-53281