Learn More
Medchemexpress LLC HY-103371 100mg Medchemexpress, DCPIB CAS:82749-70-0 Purity:>98%
SDP
Catalog No. 501968091
Encompass_Preferred
Medchemexpress, HY-103371 100mg DCPIB CAS:82749-70-0 DCPIB is a selective, reversible and potent inhibitor of volume-regulated anion channels (VRAC), voltage-dependently activates potassium channels TREK1 and TRAAK, inhibits TRESK, TASK1 and TASK3 (IC50s, 0.14, 0.95, 50.72 μM, respectively). DCPIB is also a selective blocker of swelling-induced chloride current (ICl,swell), with an IC50 of 4.1 μM in CPAE cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8091 Supplier Medchemexpress LLC Supplier No. HY103371100MG
Medchemexpress, HY-103371 100mg DCPIB CAS:82749-70-0 DCPIB is a selective, reversible and potent inhibitor of volume-regulated anion channels (VRAC), voltage-dependently activates potassium channels TREK1 and TRAAK, inhibits TRESK, TASK1 and TASK3 (IC50s, 0.14, 0.95, 50.72 μM, respectively). DCPIB is also a selective blocker of swelling-induced chloride current (ICl,swell), with an IC50 of 4.1 μM in CPAE cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.