Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dcpt1061 hydrochloride | 2289726-31-2 | 99.9% | 375.72 | 1 ML
SDP

Catalog No. 5000392887
Encompass_Preferred

DCPT1061 hydrochloride exhibits a potent inhibitory effect on PRMT1, PRMT6, and PRMT8 in vitro, with minimal inhibitory effects on epigenetic enzymes such as PRMT3, PRMT4, and PRMT5. It also possesses antitumor properties.

  • Solid appearance
  • Color is white to off-white
  • Ships at room temperature in continental US
  • Store at -20°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 50 mg/mL (133.08 mM)

Catalog No. 50-003-92887 Supplier Medchemexpress LLC Supplier No. HY162003A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DCPT1061 hydrochloride exhibits a potent inhibitory effect on PRMT1, PRMT6, and PRMT8 in vitro, with minimal inhibitory effects on epigenetic enzymes such as PRMT3, PRMT4, and PRMT5. It also possesses antitumor properties.

  • Solid appearance
  • Color is white to off-white
  • Ships at room temperature in continental US
  • Store at -20°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 50 mg/mL (133.08 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.