Learn More
Medchemexpress LLC DCPT1061 hydrochloride | 2289726-31-2 | 99.9% | 375.72 | 5 MG
SDP
Catalog No. 5000392595
Encompass_Preferred
DCPT1061 hydrochloride is a compound that exhibits a strong inhibitory effect on PRMT1, PRMT6, and PRMT8 in vitro, with little inhibitory effect on epigenetic enzymes such as PRMT3, PRMT4, and PRMT5. It also demonstrates antitumor effects.
  • Selective inhibition: Strongly inhibits PRMT1, PRMT6, and PRMT8.
  • Minimal off-target effects: Little inhibitory effect on PRMT3, PRMT4, and PRMT5.
  • Antitumor properties: Exhibits antitumor effects.
  • Research use: For research use only.
Catalog No. 50-003-92595 Supplier Medchemexpress LLC Supplier No. HY162003A5MG
DCPT1061 hydrochloride is a compound that exhibits a strong inhibitory effect on PRMT1, PRMT6, and PRMT8 in vitro, with little inhibitory effect on epigenetic enzymes such as PRMT3, PRMT4, and PRMT5. It also demonstrates antitumor effects.
  • Selective inhibition: Strongly inhibits PRMT1, PRMT6, and PRMT8.
  • Minimal off-target effects: Little inhibitory effect on PRMT3, PRMT4, and PRMT5.
  • Antitumor properties: Exhibits antitumor effects.
  • Research use: For research use only.