Learn More
Bioworld DCR Basal Salt Mixture, 25 L
Catalog No. 50254740
Encompass_Preferred
DCR Basal Salt MixtureContains Macronutrients and Micronutrients as described by Gupta & Durzan (1985)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.64 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-740 Supplier Bioworld Supplier No. 306300883
DCR Basal Salt MixtureContains Macronutrients and Micronutrients as described by Gupta & Durzan (1985)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.64 g/LpH: 3.5–4.5Soluble in water