Learn More
Sino Biological DcR1/TNFRSF10C Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10415T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DcR1/TNFRSF10C (rh DcR1/TNFRSF10C; Catalog#10415-H08H; NP_003832.2; Met1-Pro235). DcR1/TNFRSF10C specific IgG was purified by Human DcR1/TNFRSF10C affinity chromatography.
Catalog No. 50-224-8507