Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci DcR2 Peptide
SDP

Catalog No. 16918699
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-918-699 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-918-699 Supplier ProSci Supplier No. 2021P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

DcR2 peptide is used for blocking the activity of DcR2 antibody.

Specifications

Gene ID (Entrez) 8793
Species Human
Accession Number Q9UBN6
Common Name tumor necrosis factor receptor superfamily, member 10 d, decoy with truncated death domain
Concentration 200 μg/mL
Content And Storage Store DcR2 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol TNFRSF10D
Quantity 0.05 mg
Sequence 15 amino acids near the intracellular domain of human DcR2.
Synonym DCR2, CD264, TRUNDD, TRAILR4, TRAIL-R4, DCR2, UNQ251/PRO288, Tumor necrosis factor receptor superfamily member 10 D, Decoy receptor 2, DcR2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.