Learn More
ACROBiosystems ACROBiosystems Human DcR3 / TNFRSF6B Protein, Fc Tag
Supplier:  ACROBiosystems TNBH52551MG
Encompass_Preferred
Recombinant Protein;1MG;Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession # O95407-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.2 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) at 5 μg/mL (100 μL/well) can bind Human DcR3, Fc Tag (Cat. No. TNB-H5255) with a linear range of 5-20 ng/mL (QC tested).
Catalog No. 50-195-7479