Learn More
Enzo Life Sciences DcR3 (human) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522055C050
Encompass_Preferred
Alternative name: TNFRSF 6B, Decoy receptor 3. Concentration: 1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human DcR3 (decoy receptor 3) (aa 1-246) is fused to the Fc portion of human IgG1. Specificity: Binds human LIGHT. Does not bind human FasL. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9394