Learn More
Cayman Chemical DCSIGN1CD209 EXrAClular 100ug
SDP
Catalog No. 502915814
Encompass_Preferred
Source: Recombinant N-terminal human IgG1 Fc-tagged DC-SIGN1 extracellular domain expressed in HEK293 cells • Amino acids: 62-404 • MW: 65.8 kDa
Catalog No. 50-291-5814 Supplier Cayman Chemical Supplier No. 38060100 UG
Source: Recombinant N-terminal human IgG1 Fc-tagged DC-SIGN1 extracellular domain expressed in HEK293 cells • Amino acids: 62-404 • MW: 65.8 kDa