Learn More
Biomatik Corporation Dcstamp Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11909
Encompass_Preferred
Dcstamp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to DCSTAMP. The Dcstamp Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dendritic Cell-Specific Transmembrane Protein (119-209Aa). Applications: ELISA, IHC, IF. Target Synonyms: DCSTAMP. Accession Number: Q9H295. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6586