Learn More
ABclonal Technology DCSTAMP Rabbit pAb
SDP
Catalog No. 5000105058
Encompass_Preferred
This gene encodes a seven-pass transmembrane protein that is primarily expressed in dendritic cells. The encoded protein is involved in a range of immunological functions carried out by dendritic cells. This protein plays a role in osteoclastogenesis and myeloid differentiation. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-05058 Supplier ABclonal Technology Supplier No. A14630500UL
This gene encodes a seven-pass transmembrane protein that is primarily expressed in dendritic cells. The encoded protein is involved in a range of immunological functions carried out by dendritic cells. This protein plays a role in osteoclastogenesis and myeloid differentiation. Alternate splicing results in multiple transcript variants.