Learn More
Adam Equipment Adam Equipment DCT 5000 Dune Compact Portable Balance, 5kg Capacity, 2g Readability
Catalog No. NC2021399
Encompass_Preferred
Adam Equipment’s Dune balances feature basic weighing at unbeatable value. Dune has all of the necessary features for quick and easy weighing. With its advanced overload protection to prevent damage to internal components, splash-proof keypad, and sturdy plastic construction throughout, Dune delivers reliable readings in the lab or in the field. Simple to use with only 3 buttons; on/off, unit, and tare, the positive push button feel provides trouble free operation, and weighing units can be easily changed between grams, ounces and pounds. Use either the included AC adapter or 6 x AA batteries. An auto-off function saves battery life and helps reduce power consumption. Measures 5.75 inches wide x 8.25 deep x 1.6 tall.
Catalog No. NC2021399 Supplier Adam Equipment Supplier No. DCT 5000
Adam Equipment’s Dune balances feature basic weighing at unbeatable value. Dune has all of the necessary features for quick and easy weighing. With its advanced overload protection to prevent damage to internal components, splash-proof keypad, and sturdy plastic construction throughout, Dune delivers reliable readings in the lab or in the field. Simple to use with only 3 buttons; on/off, unit, and tare, the positive push button feel provides trouble free operation, and weighing units can be easily changed between grams, ounces and pounds. Use either the included AC adapter or 6 x AA batteries. An auto-off function saves battery life and helps reduce power consumption. Measures 5.75 inches wide x 8.25 deep x 1.6 tall.