Learn More
Aviva Systems Biology Corp Dct Antibody - N-terminal region (ARP45192_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45192_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dct. It was validated on Western Blot. . Alternate names: DT, TRP-2, TRP2, Tyrp-2, Tyrp2, slaty, sltApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-285-187