Learn More
Aviva Systems Biology Corp Dct Antibody - middle region (ARP45193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dct. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: DT, TRP-2, TRP2, Tyrp-2, Tyrp2, slaty, sltApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-188