Learn More
Aviva Systems Biology Corp DCTN2 Antibody - N-terminal region (ARP58450_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58450_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCTN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DCTN2. Alternate names: DCTN50, DYNAMITIN, RBP50ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62311