Learn More
Aviva Systems Biology Corp Dctn4 Antibody - C-terminal region (ARP56857_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56857_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dctn4. It was validated on Western Blot. . Alternate names: 1110001K06Rik, 4930547K17Rik, C130039E17Rik, p62ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-224