Learn More
Zymo Research Corporation dCTP (100mM)
SDP
Catalog No. 501957635
Encompass
dCTP is guaranteed for 12 months at -20°C.Avoid multiple freeze/thaw cycles. For long-term storage, aliquoting is recommended with storage at -80°C.Highlights:- Ultra high purity dCTP; >99% trisphosphate by HPLC- Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases.- Free of endo-, exodeoxyribonuclease, ribonuclease,phosphatase and nicking activities
Catalog No. 50-195-7635 Supplier Zymo Research Corporation Supplier No. D1020
dCTP is guaranteed for 12 months at -20°C.Avoid multiple freeze/thaw cycles. For long-term storage, aliquoting is recommended with storage at -80°C.Highlights:- Ultra high purity dCTP; >99% trisphosphate by HPLC- Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases.- Free of endo-, exodeoxyribonuclease, ribonuclease,phosphatase and nicking activities