Learn More
Revvity Health Sciences Inc dCTP, [α-32P]- 800Ci/mmol 10mCi/ml , 250 µCi
Catalog No. 509050824
Encompass
This item is radioactive material and cannot be ordered online
dCTP, [α-32P]- 800Ci/mmol 10mCi/ml , 250 µCi
Catalog No. 50-905-0824 Supplier Revvity Health Sciences Inc Supplier No. BLU013A250UC
This item is radioactive material and cannot be ordered online
dCTP, [α-32P]- 800Ci/mmol 10mCi/ml , 250 µCi