Learn More
Aviva Systems Biology Corp DCUN1D1 Antibody - middle region (ARP42571_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42571_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCUN1D1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DCUN1D1. Alternate names: DCUN1L1, RP42, SCCRO, SCRO, Tes3, DCNL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58270