Learn More
Aviva Systems Biology Corp DCUN1D1 Antibody - N-terminal region (ARP42570_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42570_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCUN1D1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human DCUN1D1. Alternate names: DCUN1L1, RP42, SCRO, Tes3, DCNL1, SCCROApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58269