Learn More
Aviva Systems Biology Corp DCUN1D3 Antibody - C-terminal region (ARP53231_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53231_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCUN1D3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DCUN1D3. Alternate names: 44M2.4, DKFZp686O0290, FLJ41725, MGC48972ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53704