Learn More
Medchemexpress LLC HY-19717 5mg Medchemexpress, DCVC CAS:13419-46-0 Purity:>98%
SDP
Catalog No. 502082166
Encompass_Preferred
Medchemexpress, HY-19717 5mg DCVC CAS:13419-46-0 DCVC (S-[(1E)-1,2-dichloroethenyl]--L-cysteine) is a bioactive metabolite of trichloroethylene (TCE). DCVC inhibits pathogen-stimulated pro-inflammatory cytokines IL-1β, IL-8, and TNF-α release from tissue cultures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2166 Supplier Medchemexpress LLC Supplier No. HY197175MG
Medchemexpress, HY-19717 5mg DCVC CAS:13419-46-0 DCVC (S-[(1E)-1,2-dichloroethenyl]--L-cysteine) is a bioactive metabolite of trichloroethylene (TCE). DCVC inhibits pathogen-stimulated pro-inflammatory cytokines IL-1β, IL-8, and TNF-α release from tissue cultures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.