Learn More
Aviva Systems Biology Corp DCX Antibody - C-terminal region (ARP41332_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41332_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DCX. Alternate names: DBCN, DC, LISX, SCLH, XLISApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig
Catalog No. 50-014-57805