Learn More
Aviva Systems Biology Corp DCXR Antibody - middle region (ARP42462_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42462_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCXR. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human DCXR. Alternate names: DCR, HCRII, KIDCR, P34H, XR, HCR2, SDR20C1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58234