Learn More
Medchemexpress LLC DCZ0415 | 2242470-43-3 | 99.8% | 200 MG
SDP
Catalog No. 5000341088
Encompass_Preferred
DCZ0415 is a potent TRIP13 inhibitor that impairs nonhomologous end joining repair and inhibits NF-κB activity. It induces anti-myeloma activity in vitro, in vivo, and in primary cells derived from drug-resistant myeloma patients.
  • Significantly decreases colony formation
  • Inhibits cell proliferation
  • Decreases viability in MM cells
  • Induces apoptotic cell death
  • Causes accumulation of MM cells in G0/G1 phase
  • Decreases protein levels of phosphorylated iκBα and NF-κB
  • Exerts cytotoxic effects by inhibiting DNA synthesis
  • Reduces growth of MM cells-induced tumors in immune-deficient mice
Catalog No. 50-003-41088 Supplier Medchemexpress LLC Supplier No. HY130603200MG
DCZ0415 is a potent TRIP13 inhibitor that impairs nonhomologous end joining repair and inhibits NF-κB activity. It induces anti-myeloma activity in vitro, in vivo, and in primary cells derived from drug-resistant myeloma patients.
  • Significantly decreases colony formation
  • Inhibits cell proliferation
  • Decreases viability in MM cells
  • Induces apoptotic cell death
  • Causes accumulation of MM cells in G0/G1 phase
  • Decreases protein levels of phosphorylated iκBα and NF-κB
  • Exerts cytotoxic effects by inhibiting DNA synthesis
  • Reduces growth of MM cells-induced tumors in immune-deficient mice